Assay Details
| Gene Symbol: Tmed5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028876.2 | NP_083152.1 | 3 - 4 | 638 | 69 | |
| GenBank mRNA | AK014490.1 | - | 3 - 4 | 639 | 69 | |
| AK033463.1 | - | 3 - 4 | 566 | 69 | ||
| AK170036.1 | - | 3 - 4 | 598 | 69 | ||
| BC020076.1 | - | 3 - 4 | 600 | 3985782 | 69 | |
| BC132023.1 | - | 3 - 4 | 600 | 40130300 | 69 | |
| BC138226.1 | - | 3 - 4 | 600 | 8861245 | 69 | |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0100006245.rn
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970433 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970435 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970429 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970432 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970428 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970430 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970434 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105346 | AK078317.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105362 | AK081097.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105362 | AK047805.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105362 | AK039727.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105362 | AK014490.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105362 | AK033463.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1983544 | AK085846.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1983544 | AK047805.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1983544 | AK014490.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1983544 | NM_028876.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105359 | AK140979.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105366 | AK140979.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105351 | AK042931.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 141120 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 606175 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 322336 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 841897 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 34503 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 221922 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 171722 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107662 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2089577 | AK172643.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105359 | AK140979.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1983544 | AK085846.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1983544 | AK047805.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1983544 | AK014490.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1983544 | NM_028876.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105351 | AK042931.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105346 | AK078317.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105366 | AK140979.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105362 | AK081097.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105362 | AK047805.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105362 | AK039727.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105362 | AK014490.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105362 | AK033463.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970404 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970428 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05688905 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01183464 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01183485 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107662 | - | - |
| Whole Mouse Genome G4122A | A_51_P367434 | AK085846.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P367434 | AK081097.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P367434 | AK047805.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P367434 | AK039727.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P367434 | AK014490.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108425 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107662 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04148369 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04148375 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04148393 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04148386 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970429 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970432 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970434 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970430 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970433 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970435 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04148386 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970404 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970408 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970395 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970432 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970392 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04225589 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970399 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04373348 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970418 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970430 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970424 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970406 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970421 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04993378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04148375 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04148373 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970411 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970386 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10532161 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532162 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532159 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532158 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532160 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532163 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424574_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1458962_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1445343_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431789_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424573_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10532157 | - | - |
| Mouse Genome 430 2.0 Array | 1445343_at | - | - |
| Mouse Genome 430 2.0 Array | 1424574_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424573_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431789_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1424574_at | - | - |
| Mouse Genome 430 2.0 Array | 1458962_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424573_at | - | - |
| Mouse Genome 430 2.0 Array | 1445343_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431789_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1458962_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73130_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73130_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500025998.mm.1 | NM_028876.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025998.mm.1 | NM_028876.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025998.mm.1 | TC0500002940.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500025994.mm.1 | NM_028876.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025994.mm.1 | NM_028876.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025994.mm.1 | TC0500002940.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500026008.mm.1 | NM_028876.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026008.mm.1 | NM_028876.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026008.mm.1 | TC0500002940.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500025996.mm.1 | NM_028876.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025996.mm.1 | NM_028876.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025996.mm.1 | TC0500002940.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500025999.mm.1 | NM_028876.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025999.mm.1 | NM_028876.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025999.mm.1 | TC0500002940.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500026002.mm.1 | NM_028876.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026002.mm.1 | NM_028876.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026002.mm.1 | TC0500002940.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500025995.mm.1 | NM_028876.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025995.mm.1 | NM_028876.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025995.mm.1 | TC0500002940.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500026003.mm.1 | NM_028876.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026003.mm.1 | NM_028876.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026003.mm.1 | TC0500002940.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500026012.mm.1 | NM_028876.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026012.mm.1 | NM_028876.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026012.mm.1 | TC0500002940.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500025997.mm.1 | NM_028876.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025997.mm.1 | NM_028876.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500025997.mm.1 | TC0500002940.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500026001.mm.1 | NM_028876.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026001.mm.1 | NM_028876.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026001.mm.1 | TC0500002940.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500026000.mm.1 | NM_028876.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026000.mm.1 | NM_028876.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026000.mm.1 | TC0500002940.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500026005.mm.1 | NM_028876.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026005.mm.1 | NM_028876.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026005.mm.1 | TC0500002940.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500026011.mm.1 | NM_028876.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026011.mm.1 | NM_028876.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026011.mm.1 | TC0500002940.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424573_at | - | - |
| Mouse Expression Set 430 | 1424574_PM_at | - | - |
| Mouse Expression Set 430 | 1424574_at | - | - |
| Mouse Expression Set 430 | 1424573_PM_at | - | - |
| Mouse Expression Set 430 | 1431789_PM_s_at | - | - |
| Mouse Expression Set 430 | 1445343_at | - | - |
| Mouse Expression Set 430 | 1458962_PM_at | - | - |
| Mouse Expression Set 430 | 1445343_PM_at | - | - |
| Mouse Expression Set 430 | 1458962_at | - | - |
| Mouse Expression Set 430 | 1431789_s_at | - | - |
| Murine Genome U74v2 Set | 133768_at | - | - |
| Murine Genome U74v2 Set | 133768_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424574_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424573_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424573_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424574_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445343_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458962_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445343_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458962_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 331975 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1090707 | - | - |
| MouseWG-6 v2 Expression BeadChip | 100050066 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1090707 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101190193 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 100050066 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001090707 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2713557 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1250027 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2713557 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1251790 | - | - |
| mouse-6 v1.1 (Illumina) | scl0073130.1_84-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101190193 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100050066 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1090707 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0073130.1_84-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1090707 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100050066 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101190193 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2713557 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1250027 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2713557 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2564969 | - | - |

Tmed5
Genomic Map