Assay Details
| Gene Symbol: Entpd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172117.5 | NP_742115.2 | 4 - 5 | 701 | 69 | |
| GenBank mRNA | AK030789.1 | - | 4 - 5 | 702 | 69 | |
| BC038126.1 | - | 4 - 5 | 690 | 1348160 | 69 | |
More Information
Set Membership: |
|

Entpd6
Genomic Map