Assay Details
| Gene Symbol: Zmynd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252584.2 | NP_001239513.1 | 4 - 5 | 591 | 100 | |
| NM_001252585.1 | NP_001239514.1 | 3 - 4 | 483 | 100 | ||
| NM_001252587.2 | NP_001239516.1 | 5 - 6 | 603 | 100 | ||
| NM_001281926.1 | NP_001268855.1 | 4 - 5 | 490 | 100 | ||
| NM_001291158.1 | NP_001278087.1 | 5 - 6 | 2600 | 100 | ||
| NM_027230.5 | NP_081506.3 | 5 - 6 | 678 | 100 | ||
| GenBank mRNA | AB093284.2 | - | 5 - 6 | 2592 | 100 | |
| AK132427.1 | - | 3 - 4 | 485 | 100 | ||
| AK147620.1 | - | 4 - 5 | 491 | 100 | ||
| AK155864.1 | - | 4 - 5 | 396 | 100 | ||
| BC023300.1 | - | 4 - 5 | 458 | 100 | ||
| BC048186.1 | - | 5 - 6 | 485 | 6827541 | 100 | |
| BC063267.1 | - | 4 - 5 | 384 | 100 | ||
| CF537480.1 | - | 4 - 5 | 516 | 100 | ||
| CN670238.1 | - | 5 - 6 | 418 | 100 | ||
| CO428932.1 | - | 2 - 3 | 247 | 100 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zmynd8
Genomic Map