Assay Details
| Gene Symbol: Zmynd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252584.2 | NP_001239513.1 | 7 - 8 | 887 | 89 | |
| NM_001252585.1 | NP_001239514.1 | 6 - 7 | 779 | 89 | ||
| NM_001252587.2 | NP_001239516.1 | 8 - 9 | 899 | 89 | ||
| NM_001281926.1 | NP_001268855.1 | 7 - 8 | 786 | 89 | ||
| NM_001291158.1 | NP_001278087.1 | 8 - 9 | 2896 | 89 | ||
| NM_027230.5 | NP_081506.3 | 8 - 9 | 974 | 89 | ||
| GenBank mRNA | AB093284.2 | - | 8 - 9 | 2888 | 89 | |
| AK132427.1 | - | 6 - 7 | 781 | 89 | ||
| AK147620.1 | - | 7 - 8 | 787 | 89 | ||
| AK155864.1 | - | 7 - 8 | 692 | 89 | ||
| BC023300.1 | - | 7 - 8 | 754 | 89 | ||
| BC063267.1 | - | 7 - 8 | 680 | 89 | ||
| CA328656.1 | - | 4 - 5 | 404 | 89 | ||
| CK634740.1 | - | 3 - 4 | 266 | 89 | ||
| CO428932.1 | - | 5 - 6 | 543 | 89 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zmynd8
Genomic Map