Assay Details
| Gene Symbol: P2ry10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172435.3 | NP_766023.1 | 1 - 2 | 89 | 120 | |
| GenBank mRNA | AK041351.1 | - | 1 - 2 | 105 | 120 | |
| AK088234.1 | - | 1 - 2 | 90 | 120 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK041351.1,AK088234.1,NM_172435.3 transcripts.
Set Membership: |
|

P2ry10
Genomic Map