Assay Details
| Gene Symbol: Thsd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040426.3 | NP_001035516.2 | 9 - 10 | 1812 | 99 | |
| NM_172444.4 | NP_766032.1 | 3 - 4 | 620 | 99 | ||
| GenBank mRNA | AK045414.1 | - | 3 - 4 | 536 | 99 | |
| AK138976.1 | - | 9 - 10 | 1813 | 99 | ||
| BC139329.1 | - | 3 - 4 | 513 | 8862351 | 99 | |
| CV559893.1 | - | 3 - 4 | 620 | 99 | ||
More Information
Set Membership: |
|

Thsd4
Genomic Map