Assay Details
| Gene Symbol: Thsd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040426.3 | NP_001035516.2 | 11 - 12 | 2181 | 88 | |
| NM_172444.4 | NP_766032.1 | 5 - 6 | 989 | 88 | ||
| GenBank mRNA | AK045414.1 | - | 5 - 6 | 905 | 88 | |
| AK138976.1 | - | 11 - 12 | 2182 | 88 | ||
| BC139329.1 | - | 5 - 6 | 882 | 8862351 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Thsd4
Genomic Map