Assay Details
| Gene Symbol: Thsd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040426.3 | NP_001035516.2 | 16 - 17 | 3046 | 70 | |
| NM_172444.4 | NP_766032.1 | 10 - 11 | 1854 | 70 | ||
| GenBank mRNA | AK045414.1 | - | 10 - 11 | 1770 | 70 | |
| AK138976.1 | - | 16 - 17 | 3047 | 70 | ||
| BC139329.1 | - | 10 - 11 | 1747 | 8862351 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Thsd4
Genomic Map