Assay Details
| Gene Symbol: Hmgcs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008256.4 | NP_032282.2 | 3 - 4 | 741 | 80 | |
| GenBank mRNA | AK004865.1 | - | 3 - 4 | 739 | 80 | |
| AK004902.1 | - | 3 - 4 | 737 | 80 | ||
| BC014714.1 | - | 3 - 4 | 727 | 4216637 | 80 | |
| BC024744.1 | - | 3 - 4 | 725 | 5123821 | 80 | |
More Information
Set Membership: |
|

Hmgcs2
Genomic Map