Assay Details
| Gene Symbol: Medag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027519.3 | NP_081795.1 | 1 - 2 | 445 | 111 | |
| GenBank mRNA | AB055406.1 | - | 1 - 2 | 477 | 111 | |
| AK018128.1 | - | 1 - 2 | 261 | 111 | ||
| AK036605.1 | - | 1 - 2 | 446 | 111 | ||
| AK042557.1 | - | 1 - 2 | 158 | 111 | ||
| AK047657.1 | - | 1 - 2 | 502 | 111 | ||
| AK053969.1 | - | 1 - 2 | 479 | 111 | ||
| AK086092.1 | - | 1 - 2 | 454 | 111 | ||
| AK138268.1 | - | 1 - 2 | 144 | 111 | ||
| AK160904.1 | - | 1 - 2 | 452 | 111 | ||
| AK161526.1 | - | 1 - 2 | 453 | 112 | ||
| AK162725.1 | - | 1 - 2 | 446 | 111 | ||
| AK163617.1 | - | 1 - 2 | 144 | 111 | ||
| BC026658.1 | - | 1 - 2 | 208 | 111 | ||
| BC116246.1 | - | 1 - 2 | 367 | 40093530 | 111 | |
| BC116247.1 | - | 1 - 2 | 365 | 40093536 | 111 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK042557.1,AK138268.1,AK163617.1 transcripts.
Set Membership: |
|

Medag
Genomic Map