Assay Details
| Gene Symbol: Ndc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028355.3 | NP_082631.1 | 7 - 8 | 1064 | 60 | |
| GenBank mRNA | AK013409.1 | - | 7 - 8 | 801 | 60 | |
| AK144734.1 | - | 7 - 8 | 1065 | 60 | ||
| BC021337.1 | - | 7 - 8 | 819 | 5042118 | 60 | |
| DQ141695.1 | - | 7 - 8 | 759 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ndc1
Genomic Map