Assay Details
| Gene Symbol: Olfr1056 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147018.2 | NP_667229.2 | 1 | 190 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2074698 | NM_147018.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2074698 | BC145903.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109154 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2074698 | NM_147018.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2074698 | BC145903.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P206907 | NM_147018.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P206907 | BC145903.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109154 | - | - |
| Whole Mouse Genome G4122A | A_51_P206907 | NM_147018.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P206907 | BC145903.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126766 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401928 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01648005 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401932 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401904 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01681832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401935 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401910 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401921 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01648029 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01284515 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01920657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401934 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401896 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401938 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401892 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01690400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401915 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401893 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10484591 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10484590 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200033756.mm.1 | NM_147018.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200033756.mm.1 | NM_147018.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200033756.mm.1 | TC0200003931.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200033755.mm.1 | NM_147018.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200033755.mm.1 | NM_147018.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200033755.mm.1 | TC0200003931.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 694358 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4050072 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004050072 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1250553 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1250553 | - | - |
| mouse-6 v1.1 (Illumina) | scl4338.1.1_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4050072 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl4338.1.1_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4050072 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2982525 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1250553 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2982525 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl4338.1.1_1-S | - | - |

Olfr1056
Genomic Map