Assay Details
| Gene Symbol: Fuk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172283.3 | NP_758487.2 | 15 - 16 | 1871 | 54 | |
| NR_052009.1 | 15 - 16 | 1969 | 54 | |||
| GenBank mRNA | AJ297482.4 | - | 15 - 16 | 1842 | 54 | |
| AJ534942.1 | - | 15 - 16 | 1842 | 54 | ||
| AK049153.1 | - | 4 - 5 | 700 | 54 | ||
| AK164340.1 | - | 15 - 16 | 1938 | 54 | ||
| AY223865.1 | - | 15 - 16 | 1842 | 54 | ||
| BC037698.1 | - | 14 - 15 | 1820 | 4190449 | 54 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26165490 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26165491 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26165492 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26165493 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017183 | AK164340.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017183 | AJ534942.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017183 | AJ297482.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017183 | NM_181666.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2031786 | BC037698.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2031786 | AY223865.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2031786 | AK164340.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2031786 | AJ534942.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2031786 | AJ297482.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 968819 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 971049 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 538415 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 386442 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 674957 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 520511 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 751225 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 748354 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 683888 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 296805 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 936486 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 1508 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 128716 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166374 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129746 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017183 | AK164340.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017183 | AJ534942.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017183 | AJ297482.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017183 | NM_181666.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2031786 | BC037698.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2031786 | AY223865.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2031786 | AK164340.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2031786 | AJ534942.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2031786 | AJ297482.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P464361 | BC037698.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P464361 | AK131165.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P464361 | AY223865.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P464361 | AK164340.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P464361 | AK049153.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26165490 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26165486 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26165506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26165509 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26165500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26165484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26165498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26165503 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26165496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26165493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26165485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26165497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26165494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26165491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26165487 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26165481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26165507 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26165488 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129746 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166374 | - | - |
| Whole Mouse Genome G4122A | A_51_P464361 | BC037698.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P464361 | AK131165.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P464361 | AY223865.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P464361 | AK164340.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P464361 | AK049153.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100675 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129746 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166374 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07294375 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06978893 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06978344 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07294374 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07081908 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26165491 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26165493 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26165492 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26165490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26165315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26165511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26165485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06887497 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26165463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26165507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26165481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26165508 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26165516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07294379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26165493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26165506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07057215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26165467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07418044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26165473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26165487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06978341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06955948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07143205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26165468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07421538 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26165466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26165494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06714555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26165510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26165498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26165514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26165465 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26165484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26165479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06714560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26165490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26165496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26165503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26165495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07097482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26165464 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10581728 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581722 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581716 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581706 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581707 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581711 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581714 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581727 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581718 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581721 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581725 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581723 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581726 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581720 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581724 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581709 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581713 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581704 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581703 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581705 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581710 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581717 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581708 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581715 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581719 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581712 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437986_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437337_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454634_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10581702 | - | - |
| Mouse Genome 430 2.0 Array | 1437337_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1437986_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1437337_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1454634_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1437986_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1454634_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 234730_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 234730_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800022537.mm.1 | NM_172283.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800022537.mm.1 | NR_052009.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800022537.mm.1 | TC0800002931.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1437986_PM_x_at | - | - |
| Mouse Expression Set 430 | 1437337_PM_x_at | - | - |
| Mouse Expression Set 430 | 1454634_PM_at | - | - |
| Mouse Expression Set 430 | 1454634_at | - | - |
| Mouse Expression Set 430 | 1437337_x_at | - | - |
| Mouse Expression Set 430 | 1437986_x_at | - | - |
| Murine Genome U74v2 Set | 170970_i_at | - | - |
| Murine Genome U74v2 Set | 170071_r_at | - | - |
| Murine Genome U74v2 Set | 170071_PM_r_at | - | - |
| Murine Genome U74v2 Set | 170970_PM_i_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437986_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437337_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454634_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437337_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437986_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454634_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 341883 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5050575 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5050575 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005050575 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2641632 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2641632 | - | - |
| mouse-6 v1.1 (Illumina) | scl34329.24.1_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5050575 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl34329.24.1_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5050575 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2641632 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3120565 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3120565 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2641632 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl34329.24.1_1-S | - | - |

Fuk
Genomic Map