Assay Details
| Gene Symbol: Fuk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172283.3 | NP_758487.2 | 19 - 20 | 2612 | 94 | |
| NR_052009.1 | 19 - 20 | 2710 | 94 | |||
| GenBank mRNA | AJ297482.4 | - | 19 - 20 | 2583 | 94 | |
| AJ534942.1 | - | 19 - 20 | 2583 | 94 | ||
| AK049153.1 | - | 8 - 9 | 1441 | 94 | ||
| AK131165.1 | - | 8 - 9 | 3962 | 94 | ||
| AK161763.1 | - | 19 - 20 | 2716 | 94 | ||
| AY223865.1 | - | 19 - 20 | 2583 | 94 | ||
| BC037698.1 | - | 18 - 19 | 2561 | 4190449 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fuk
Genomic Map