| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900020686.mm.1 |
NM_172446.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020686.mm.1 |
NM_172446.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020686.mm.1 |
TC0900002471.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900020685.mm.1 |
NM_001163755.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020685.mm.1 |
NM_001163755.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020685.mm.1 |
NM_001163757.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020685.mm.1 |
NM_001163757.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020685.mm.1 |
NM_001163758.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020685.mm.1 |
NM_001163758.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020685.mm.1 |
NM_172446.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020685.mm.1 |
NM_172446.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020685.mm.1 |
TC0900002471.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900020687.mm.1 |
NM_001163755.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020687.mm.1 |
NM_001163755.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020687.mm.1 |
NM_001163757.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020687.mm.1 |
NM_001163757.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020687.mm.1 |
NM_172446.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020687.mm.1 |
NM_172446.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020687.mm.1 |
TC0900002471.mm.1 |
MTA TC |