Assay Details
| Gene Symbol: Skor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163755.1 | NP_001157227.1 | 7 - 8 | 2745 | 67 | |
| NM_001163757.1 | NP_001157229.1 | 7 - 8 | 2745 | 67 | ||
| NM_001163758.1 | NP_001157230.1 | 7 - 8 | 2726 | 67 | ||
| NM_172446.3 | NP_766034.2 | 7 - 8 | 2798 | 67 | ||
| GenBank mRNA | AB185113.1 | - | 7 - 8 | 2748 | 67 | |
| AK029916.1 | - | 7 - 8 | 2729 | 67 | ||
| AK049035.1 | - | 7 - 8 | 2799 | 67 | ||
| AK134840.1 | - | 7 - 8 | 2737 | 67 | ||
| BC138063.1 | - | 7 - 8 | 2702 | 8861082 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Skor1
Genomic Map