Assay Details
| Gene Symbol: Rnf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172448.3 | NP_766036.2 | 4 - 5 | 1068 | 72 | |
| GenBank mRNA | AK028750.1 | - | 3 - 4 | 427 | 72 | |
| AK153638.1 | - | 3 - 4 | 2346 | 72 | ||
| BC011216.1 | - | 3 - 4 | 794 | 72 | ||
| BC075707.1 | - | 2 - 3 | 204 | 72 | ||
| BC131957.1 | - | 2 - 3 | 186 | 40130234 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnf43
Genomic Map