Assay Details
| Gene Symbol: Rnf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172448.3 | NP_766036.2 | 8 - 9 | 1569 | 106 | |
| GenBank mRNA | AK028750.1 | - | 7 - 8 | 928 | 106 | |
| BC075707.1 | - | 6 - 7 | 705 | 106 | ||
| BC131957.1 | - | 6 - 7 | 687 | 40130234 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnf43
Genomic Map