Assay Details
| Gene Symbol: Tmem44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172614.3 | NP_766202.3 | 2 - 3 | 339 | 99 | |
| GenBank mRNA | AK031524.1 | - | 2 - 3 | 301 | 99 | |
| AK045675.1 | - | 3 - 4 | 402 | 99 | ||
| AK048655.1 | - | 2 - 3 | 340 | 99 | ||
| AK082652.1 | - | 2 - 3 | 1574 | 99 | ||
| BC059216.1 | - | 2 - 3 | 677 | 99 | ||
| BC080307.1 | - | 3 - 4 | 235 | 99 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK045675.1,AK048655.1,AK082652.1 transcripts.
Set Membership: |
|

Tmem44
Genomic Map