Assay Details
| Gene Symbol: Tmem44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172614.3 | NP_766202.3 | 8 - 9 | 1114 | 104 | |
| GenBank mRNA | AK031524.1 | - | 7 - 8 | 3566 | 104 | |
| AK045675.1 | - | 9 - 10 | 1177 | 104 | ||
| AK048655.1 | - | 8 - 9 | 1115 | 104 | ||
| AK079084.1 | - | 3 - 4 | 440 | 104 | ||
| AK082652.1 | - | 8 - 9 | 2350 | 104 | ||
| BC059216.1 | - | 8 - 9 | 1452 | 104 | ||
| BC080307.1 | - | 9 - 10 | 1010 | 104 | ||
More Information
Set Membership: |
|

Tmem44
Genomic Map