Assay Details
| Gene Symbol: Sh3tc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172628.2 | NP_766216.2 | 5 - 6 | 575 | 139 | |
| GenBank mRNA | AK052534.1 | - | 5 - 6 | 567 | 139 | |
| AK143600.1 | - | 576 | 139 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3tc2
Genomic Map