Assay Details
| Gene Symbol: Sh3tc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172628.2 | NP_766216.2 | 6 - 7 | 769 | 82 | |
| GenBank mRNA | AK052534.1 | - | 6 - 7 | 761 | 82 | |
| AK143600.1 | - | 770 | 82 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3tc2
Genomic Map