Assay Details
| Gene Symbol: Sh3tc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172628.2 | NP_766216.2 | 10 - 11 | 1211 | 68 | |
| GenBank mRNA | AK052534.1 | - | 10 - 11 | 1203 | 68 | |
| AK143600.1 | - | 1212 | 68 | |||
| AK173317.1 | - | 2 - 3 | 144 | 68 | ||
| BC076633.1 | - | 3 - 4 | 286 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3tc2
Genomic Map