Assay Details
| Gene Symbol: Sh3tc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172628.2 | NP_766216.2 | 14 - 15 | 3374 | 62 | |
| GenBank mRNA | AK052534.1 | - | 14 - 15 | 3326 | 62 | |
| AK143600.1 | - | 3375 | 62 | |||
| AK173317.1 | - | 6 - 7 | 2307 | 62 | ||
| BC037000.1 | - | 2 - 3 | 216 | 62 | ||
| BC049856.1 | - | 1 - 2 | 54 | 62 | ||
| BC076633.1 | - | 7 - 8 | 2449 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3tc2
Genomic Map