Assay Details
| Gene Symbol: Sh3tc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172628.2 | NP_766216.2 | 15 - 16 | 3525 | 53 | |
| GenBank mRNA | AK052534.1 | - | 15 - 16 | 3477 | 53 | |
| AK143600.1 | - | 3526 | 53 | |||
| AK173317.1 | - | 7 - 8 | 2458 | 53 | ||
| BC037000.1 | - | 3 - 4 | 367 | 53 | ||
| BC049856.1 | - | 2 - 3 | 205 | 53 | ||
| BC076633.1 | - | 8 - 9 | 2600 | 53 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3tc2
Genomic Map