Assay Details
| Gene Symbol: Dars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172644.3 | NP_766232.1 | 8 - 9 | 1281 | 80 | |
| GenBank mRNA | AK030952.1 | - | 8 - 9 | 1270 | 80 | |
| AK033634.1 | - | 7 - 8 | 2104 | 80 | ||
| AK037337.1 | - | 8 - 9 | 1300 | 80 | ||
| AK048454.1 | - | 8 - 9 | 1258 | 80 | ||
| AK138809.1 | - | 8 - 9 | 1282 | 80 | ||
| AK154133.1 | - | 8 - 9 | 1309 | 80 | ||
| AK154348.1 | - | 8 - 9 | 1309 | 80 | ||
| AK164885.1 | - | 8 - 9 | 1251 | 80 | ||
| AK216242.1 | - | 3 - 4 | 177 | 80 | ||
| BC057943.1 | - | 8 - 9 | 1305 | 80 | ||
| BC076606.1 | - | 8 - 9 | 1273 | 30625515 | 80 | |
| BC089191.1 | - | 8 - 9 | 1255 | 30473852 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dars2
Genomic Map