Assay Details
| Gene Symbol: N28178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172690.2 | NP_766278.1 | 3 - 4 | 339 | 64 | |
| GenBank mRNA | AK038528.1 | - | 2 - 3 | 193 | 64 | |
| AK046283.1 | - | 3 - 4 | 330 | 64 | ||
| BC057092.1 | - | 3 - 4 | 339 | 5703966 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

N28178
Genomic Map