| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400002700.mm.1 |
NM_172690.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400002700.mm.1 |
NM_172690.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400002700.mm.1 |
TC0400000425.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400002701.mm.1 |
NM_172690.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400002701.mm.1 |
NM_172690.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400002701.mm.1 |
TC0400000425.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400002704.mm.1 |
NM_172690.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400002704.mm.1 |
NM_172690.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400002704.mm.1 |
TC0400000425.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400002703.mm.1 |
NM_172690.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400002703.mm.1 |
NM_172690.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400002703.mm.1 |
TC0400000425.mm.1 |
MTA TC |