Assay Details
| Gene Symbol: Nxt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161430.2 | NP_001154902.1 | 2 - 3 | 166 | 109 | |
| NM_001290532.1 | NP_001277461.1 | 2 - 3 | 251 | 109 | ||
| NM_172782.4 | NP_766370.2 | 1 - 2 | 203 | 109 | ||
| GenBank mRNA | AK032102.1 | - | 1 - 2 | 174 | 109 | |
| AK144349.1 | - | 1 - 2 | 187 | 109 | ||
| BC064727.1 | - | 1 - 2 | 178 | 109 | ||
| BC068166.1 | - | 1 - 2 | 156 | 30361776 | 109 | |
| BC094570.1 | - | 1 - 2 | 120 | 30361291 | 109 | |
| BC118503.1 | - | 1 - 2 | 131 | 40106658 | 109 | |
| CA482101.1 | - | 2 - 3 | 202 | 109 | ||
| CJ148018.1 | - | 2 - 3 | 252 | 109 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK032102.1,AK144349.1,BC068166.1,BC094570.1,BC118503.1,NM_001161430.2,NM_172782.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990041 | BC068166.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990041 | BC094570.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990041 | AK144349.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990041 | BC064727.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990041 | NM_172782.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 543544 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 568210 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923907 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 918616 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 345612 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 144187 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 127094 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141416 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157507 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990041 | BC068166.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990041 | BC094570.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990041 | AK144349.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990041 | BC064727.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990041 | NM_172782.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517904 | BC068166.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517904 | BC094570.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517904 | BC118503.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517904 | AK144349.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517904 | BC064727.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707042 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707028 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707030 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707046 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707035 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707025 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707037 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707031 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32707036 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157507 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P141416 | - | - |
| Whole Mouse Genome G4122A | A_51_P517904 | BC068166.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517904 | BC094570.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517904 | BC118503.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517904 | AK144349.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517904 | BC064727.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130955 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P141416 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157507 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15093626 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15093622 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15093624 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15093618 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15093627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32707115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707061 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14607811 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707046 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14612115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707028 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707048 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707032 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707056 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707043 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14943313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707044 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707050 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707024 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707071 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707018 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707068 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32707060 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10602167 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10602171 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10602168 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10602170 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10602172 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10602169 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456599_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10602166 | - | - |
| Mouse Genome 430 2.0 Array | 1456599_at | - | - |
| Mouse Genome 430 2.0 Array | 1456599_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 237082_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 237082_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00008884.mm.1 | NM_001161430.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008884.mm.1 | TC0X00001402.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008886.mm.1 | NM_172782.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008886.mm.1 | TC0X00001402.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008893.mm.1 | NM_001161430.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008893.mm.1 | NM_172782.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008893.mm.1 | TC0X00001402.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008890.mm.1 | NM_172782.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008890.mm.1 | TC0X00001402.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008891.mm.1 | NM_001161430.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008891.mm.1 | NM_172782.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008891.mm.1 | TC0X00001402.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008895.mm.1 | NM_001161430.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008895.mm.1 | NM_172782.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008895.mm.1 | TC0X00001402.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1456599_at | - | - |
| Mouse Expression Set 430 | 1456599_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456599_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456599_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 759487 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2900008 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002900008 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1245658 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1245658 | - | - |
| mouse-6 v1.1 (Illumina) | scl0237082.4_163-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2900008 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0237082.4_163-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2900008 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1245658 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1245658 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0237082.4_163-S | - | - |

Nxt2
Genomic Map