Assay Details
| Gene Symbol: Wdr63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172864.3 | NP_766452.2 | 16 - 17 | 1988 | 99 | |
| GenBank mRNA | AK029903.1 | - | 16 - 17 | 1990 | 99 | |
| BC132106.1 | - | 11 - 12 | 1426 | 40130383 | 99 | |
| BC132108.1 | - | 11 - 12 | 1426 | 40130385 | 99 | |
| BC158119.1 | - | 16 - 17 | 2094 | 9007392 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr63
Genomic Map