Assay Details
| Gene Symbol: Wdr63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172864.3 | NP_766452.2 | 20 - 21 | 2549 | 109 | |
| GenBank mRNA | AK029903.1 | - | 20 - 21 | 2551 | 109 | |
| BC132106.1 | - | 15 - 16 | 1987 | 40130383 | 109 | |
| BC132108.1 | - | 15 - 16 | 1987 | 40130385 | 109 | |
| BC158119.1 | - | 20 - 21 | 2655 | 9007392 | 109 | |
More Information
Set Membership: |
|

Wdr63
Genomic Map