Assay Details
| Gene Symbol: Ccdc113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172914.2 | NP_766502.1 | 3 - 4 | 461 | 69 | |
| GenBank mRNA | AK077130.1 | - | 3 - 4 | 462 | 69 | |
| BC048506.1 | - | 3 - 4 | 422 | 69 | ||
| BC060957.1 | - | 3 - 4 | 459 | 6705542 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1981704 | NM_172914.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1981704 | AK077130.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 351943 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 232171 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 246761 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 641118 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 789825 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 910003 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 187378 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 828113 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 922925 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 862629 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 123473 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 556729 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 97423 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 118482 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121851 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170314 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1981704 | NM_172914.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1981704 | AK077130.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26083796 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26083826 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26083823 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26083799 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26083801 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26083818 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26083810 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26083825 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26083794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26083816 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26083807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26083802 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26083820 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26083798 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26083814 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26083812 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26083804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26083819 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26083822 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01803170 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P170314 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P121851 | - | - |
| Whole Mouse Genome G4122A | A_51_P410576 | BC060957.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P410576 | NM_172914.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P410576 | AK077130.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116201 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P170314 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P121851 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01803170 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07021464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26084000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26083781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083881 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06832222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07341170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083810 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083816 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06832223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07381626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06744563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07179268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083797 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07204631 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07382120 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06744555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083837 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083807 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083878 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083794 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083857 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083835 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083879 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083792 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083825 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07226688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07204634 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083833 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083850 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083823 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06730569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083841 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083829 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06832226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083818 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07021471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083822 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083775 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083874 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083814 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083868 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083826 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083941 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06973432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083845 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06767840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26083803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06742606 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10574370 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574373 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574368 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574366 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574374 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574369 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574367 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574372 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574371 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436285_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10574365 | - | - |
| Mouse Genome 430 2.0 Array | 1436285_at | - | - |
| Mouse Genome 430 2.0 Array | 1436285_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 244608_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 244608_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800008954.mm.1 | NM_172914.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008954.mm.1 | NM_172914.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008954.mm.1 | TC0800001137.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1436285_PM_at | - | - |
| Mouse Expression Set 430 | 1436285_at | - | - |
| Murine Genome U74v2 Set | 165769_f_at | - | - |
| Murine Genome U74v2 Set | 165769_PM_f_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436285_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436285_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 609505 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3140010 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003140010 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2610277 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2610277 | - | - |
| mouse-6 v1.1 (Illumina) | scl33459.9.1_80-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3140010 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl33459.9.1_80-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3140010 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2610277 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2610277 | - | - |

Ccdc113
Genomic Map