Assay Details
| Gene Symbol: AI118078 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172923.3 | NP_766511.1 | 2 - 3 | 182 | 110 | |
| GenBank mRNA | AK036040.1 | - | 2 - 3 | 182 | 110 | |
| BC068128.1 | - | 2 - 3 | 218 | 6835202 | 110 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK036040.1, BC068128.1, NM_172923.3 transcripts.
Set Membership: |
|

AI118078
Genomic Map