Assay Details
| Gene Symbol: AI118078 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172923.3 | NP_766511.1 | 7 - 8 | 797 | 68 | |
| GenBank mRNA | AK036040.1 | - | 7 - 8 | 797 | 68 | |
| BC068128.1 | - | 8 - 9 | 953 | 6835202 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

AI118078
Genomic Map