| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900004856.mm.1 |
NM_172923.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004856.mm.1 |
NM_172923.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004856.mm.1 |
TC0900000674.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900004859.mm.1 |
NM_172923.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004859.mm.1 |
NM_172923.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004859.mm.1 |
TC0900000674.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900004858.mm.1 |
NM_172923.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004858.mm.1 |
NM_172923.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004858.mm.1 |
TC0900000674.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900004857.mm.1 |
TC0900000674.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900004860.mm.1 |
NM_172923.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004860.mm.1 |
NM_172923.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004860.mm.1 |
TC0900000674.mm.1 |
MTA TC |