Assay Details
| Gene Symbol: Sh3rf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146299.1 | NP_001139771.1 | 7 - 8 | 1687 | 81 | |
| NM_172966.3 | NP_766554.2 | 6 - 7 | 1591 | 81 | ||
| GenBank mRNA | AK028153.1 | - | 6 - 7 | 1087 | 81 | |
| AK029168.1 | - | 6 - 7 | 1592 | 81 | ||
| AK033601.1 | - | 7 - 8 | 1731 | 81 | ||
| BC048400.1 | - | 1 - 2 | 256 | 81 | ||
| BC137955.1 | - | 7 - 8 | 1640 | 8860974 | 81 | |
| BC145060.1 | - | 7 - 8 | 1640 | 9053608 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sh3rf2
Genomic Map