| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200027322.mm.1 |
NM_001166033.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027322.mm.1 |
NM_001166033.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027322.mm.1 |
NM_172977.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027322.mm.1 |
NM_172977.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027322.mm.1 |
TC0200003238.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200027320.mm.1 |
NM_001166033.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027320.mm.1 |
NM_001166033.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027320.mm.1 |
NM_172977.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027320.mm.1 |
NM_172977.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027320.mm.1 |
TC0200003238.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200027321.mm.1 |
NM_001166033.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027321.mm.1 |
NM_001166033.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027321.mm.1 |
NM_172977.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027321.mm.1 |
NM_172977.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027321.mm.1 |
TC0200003238.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200027325.mm.1 |
NM_001166033.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027325.mm.1 |
NM_001166033.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027325.mm.1 |
NM_172977.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027325.mm.1 |
NM_172977.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027325.mm.1 |
TC0200003238.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200027329.mm.1 |
NM_001166033.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027329.mm.1 |
NM_001166033.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027329.mm.1 |
NM_172977.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027329.mm.1 |
NM_172977.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027329.mm.1 |
TC0200003238.mm.1 |
MTA TC |