Assay Details
| Gene Symbol: Zfp512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172993.3 | NP_766581.2 | 1 - 2 | 141 | 101 | |
| GenBank mRNA | AK084205.1 | - | 1 - 2 | 88 | 101 | |
| AK146353.1 | - | 1 - 2 | 105 | 101 | ||
| AK149921.1 | - | 1 - 2 | 88 | 101 | ||
| AK154918.1 | - | 1 - 2 | 88 | 101 | ||
| AK173267.1 | - | 1 - 2 | 116 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK084205.1,AK146353.1,AK149921.1,AK154918.1,NM_172993.3 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zfp512
Genomic Map