Assay Details
| Gene Symbol: Zfp512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172993.3 | NP_766581.2 | 6 - 7 | 703 | 80 | |
| GenBank mRNA | AK084205.1 | - | 6 - 7 | 650 | 80 | |
| AK146353.1 | - | 6 - 7 | 667 | 80 | ||
| AK154918.1 | - | 6 - 7 | 650 | 80 | ||
| AK173267.1 | - | 6 - 7 | 678 | 80 | ||
| BC021520.1 | - | 4 - 5 | 513 | 80 | ||
| BC066136.1 | - | 6 - 7 | 626 | 80 | ||
| BC089160.1 | - | 6 - 7 | 625 | 5390706 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp512
Genomic Map