Assay Details
| Gene Symbol: Zfp512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172993.3 | NP_766581.2 | 10 - 11 | 1247 | 80 | |
| GenBank mRNA | AK084205.1 | - | 10 - 11 | 1194 | 80 | |
| AK146353.1 | - | 10 - 11 | 1211 | 80 | ||
| AK149921.1 | - | 10 - 11 | 1194 | 80 | ||
| AK154918.1 | - | 10 - 11 | 1194 | 80 | ||
| AK173267.1 | - | 10 - 11 | 1222 | 80 | ||
| BC021520.1 | - | 8 - 9 | 1041 | 80 | ||
| BC066136.1 | - | 9 - 10 | 1000 | 80 | ||
| BC089160.1 | - | 10 - 11 | 1169 | 5390706 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp512
Genomic Map