| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100038335.mm.1 |
NM_001252067.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038335.mm.1 |
NM_001252067.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038335.mm.1 |
NM_173048.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038335.mm.1 |
NM_173048.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038335.mm.1 |
TC1100004081.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100038338.mm.1 |
NM_001252067.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038338.mm.1 |
NM_001252067.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038338.mm.1 |
NM_173048.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038338.mm.1 |
NM_173048.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038338.mm.1 |
TC1100004081.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100038337.mm.1 |
NM_001252067.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038337.mm.1 |
NM_001252067.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038337.mm.1 |
NM_173048.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038337.mm.1 |
NM_173048.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038337.mm.1 |
TC1100004081.mm.1 |
MTA TC |