Assay Details
| Gene Symbol: Zfyve16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173392.4 | NP_775568.1 | 10 - 11 | 3717 | 71 | |
| GenBank mRNA | AK122241.1 | - | 10 - 11 | 3698 | 71 | |
| BC042669.1 | - | 10 - 11 | 3715 | 4235024 | 71 | |
More Information
Set Membership: |
|

Zfyve16
Genomic Map