Assay Details
| Gene Symbol: Bmp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173404.3 | NP_775580.1 | 1 - 2 | 988 | 101 | |
| GenBank mRNA | AK033644.1 | - | 1 - 2 | 626 | 101 | |
| AK035299.1 | - | 1 - 2 | 1014 | 101 | ||
| AK035393.1 | - | 1 - 2 | 977 | 101 | ||
| AK037225.1 | - | 1 - 2 | 969 | 101 | ||
| AK040389.1 | - | 1 - 2 | 989 | 101 | ||
| BC117749.2 | - | 1 - 2 | 601 | 40090489 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bmp3
Genomic Map