Assay Details
| Gene Symbol: Gpbar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174985.1 | NP_778150.1 | 1 | 944 | 73 | |
| GenBank mRNA | AB086171.1 | - | 1 | 944 | 73 | |
| AB089308.1 | - | 1 | 944 | 73 | ||
| BC116914.1 | - | 1 | 994 | 40126233 | 73 | |
| BC119327.1 | - | 1 | 994 | 8734076 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2101441 | BC119327.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2101441 | BC116914.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2101441 | NM_174985.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2101441 | AB086171.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2101441 | AB089308.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138067 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2101441 | BC119327.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2101441 | BC116914.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2101441 | NM_174985.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2101441 | AB086171.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2101441 | AB089308.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P175311 | BC119327.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P175311 | BC116914.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P175311 | NM_174985.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P175311 | AB086171.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P175311 | AB089308.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347605 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347638 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347640 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347613 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347637 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347636 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347641 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347607 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347619 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347644 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347615 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347632 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347618 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347608 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20347617 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138067 | - | - |
| Whole Mouse Genome G4122A | A_51_P175311 | BC119327.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175311 | BC116914.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175311 | NM_174985.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175311 | AB086171.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175311 | AB089308.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113248 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138067 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00388489 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01065544 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00275693 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00388458 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01065549 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00388507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347605 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00388453 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00202938 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00202936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00275697 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347632 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00128538 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00388455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347604 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00031557 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00275701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347607 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00388507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00388508 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00275699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00202935 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00122789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01065547 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347615 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00810666 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347628 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00388458 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347606 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347605 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01065545 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347638 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00275695 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00202939 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347601 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00275700 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00388489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00202944 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10347309 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10347308 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10347307 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100005023.mm.1 | TC0100000626.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 634979 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2320452 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2320452 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002320452 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1248346 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1248346 | - | - |
| mouse-6 v1.1 (Illumina) | scl17804.1.1_157-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2320452 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl17804.1.1_157-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2320452 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1248346 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1248346 | - | - |

Gpbar1
Genomic Map