Assay Details
| Gene Symbol: Upk3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175309.4 | NP_780518.1 | 4 - 5 | 852 | 66 | |
| GenBank mRNA | AK042067.1 | - | 4 - 5 | 648 | 66 | |
| AK052525.1 | - | 4 - 5 | 854 | 66 | ||
| AK157647.1 | - | 4 - 5 | 659 | 66 | ||
| AY233463.1 | - | 4 - 5 | 646 | 66 | ||
| BC120594.1 | - | 4 - 5 | 602 | 40129517 | 66 | |
| BC137742.1 | - | 4 - 5 | 602 | 8860761 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Upk3b
Genomic Map