Assay Details
| Gene Symbol: Igfbp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008344.3 | NP_032370.2 | 3 - 4 | 956 | 91 | |
| GenBank mRNA | AW123368.1 | - | 1 - 2 | 350 | 91 | |
| BC012723.1 | - | 3 - 4 | 663 | 4224052 | 91 | |
| CT010184.1 | - | 3 - 4 | 594 | 91 | ||
| X81584.1 | - | 3 - 4 | 91 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30496259 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30496261 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30496260 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30496262 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30496258 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P139546 | NM_008344.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P139546 | AK157982.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P139546 | AW123368.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P139546 | BC012723.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P139546 | X81584.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 761204 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 243681 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 173233 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150108 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P139546 | NM_008344.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P139546 | AK157982.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P139546 | AW123368.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P139546 | BC012723.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P139546 | X81584.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P374571 | X81584.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30496282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30496265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30496260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30496271 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30496258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30496286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30496263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30496268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30496284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30496270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30496276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30496277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30496274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30496267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30496262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30496280 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03117984 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150108 | - | - |
| Whole Mouse Genome G4122A | A_51_P374571 | X81584.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105897 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139546 | NM_008344.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139546 | AK157982.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139546 | AW123368.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139546 | BC012723.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139546 | X81584.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150108 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12402455 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30496259 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30496261 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30496260 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30496262 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30496258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30496289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12133356 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496276 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12081899 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496280 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496248 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496256 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496277 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496252 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11950855 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496233 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496281 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496275 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496245 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12402472 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12236182 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496250 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496268 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496254 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12169496 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496263 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496234 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12402465 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496262 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496237 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496267 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496284 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12236181 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496243 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12230606 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496232 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30496241 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10427130 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427127 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427126 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427129 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427128 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417933_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10427125 | - | - |
| Mouse Genome 430 2.0 Array | 1417933_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417933_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16012_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16012_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500008645.mm.1 | NM_008344.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008645.mm.1 | NM_008344.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008645.mm.1 | TC1500001089.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500008647.mm.1 | NM_008344.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008647.mm.1 | NM_008344.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008647.mm.1 | TC1500001089.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500008643.mm.1 | NM_008344.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008643.mm.1 | NM_008344.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008643.mm.1 | TC1500001089.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500008644.mm.1 | NM_008344.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008644.mm.1 | NM_008344.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008644.mm.1 | TC1500001089.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500008646.mm.1 | NM_008344.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008646.mm.1 | NM_008344.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008646.mm.1 | TC1500001089.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500008648.mm.1 | NM_008344.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008648.mm.1 | NM_008344.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008648.mm.1 | TC1500001089.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417933_at | - | - |
| Mouse Expression Set 430 | 1417933_PM_at | - | - |
| Murine Genome U74v2 Set | 103904_at | - | - |
| Murine Genome U74v2 Set | 103904_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417933_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417933_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417933_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417933_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 586618 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5900253 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5900253 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005900253 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2689790 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2689790 | - | - |
| mouse-6 v1.1 (Illumina) | scl016012.5_14-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5900253 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl016012.5_14-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5900253 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2689790 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2689790 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl016012.5_14-S | - | - |

Igfbp6
Genomic Map