Assay Details
| Gene Symbol: Lap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024434.6 | NP_077754.3 | 1 - 2 | 193 | 101 | |
| GenBank mRNA | AF334160.1 | - | 1 - 2 | 154 | 101 | |
| AK005334.1 | - | 1 - 2 | 106 | 101 | ||
| AK008600.1 | - | 1 - 2 | 80 | 101 | ||
| AK010502.1 | - | 1 - 2 | 146 | 101 | ||
| AK011604.1 | - | 1 - 2 | 79 | 101 | ||
| AK089023.1 | - | 1 - 2 | 160 | 101 | ||
| AK166958.1 | - | 1 - 2 | 77 | 101 | ||
| AK169032.1 | - | 1 - 2 | 79 | 101 | ||
| BC016536.1 | - | 1 - 2 | 193 | 3488469 | 101 | |
| CT010237.1 | - | 1 - 2 | 107 | 101 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK005334.1,AK008600.1,AK011604.1,AK166958.1,AK169032.1 transcripts.
Set Membership: |
|

Lap3
Genomic Map