Assay Details
| Gene Symbol: Tex21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159532.1 | NP_001153004.1 | 7 - 8 | 1449 | 133 | |
| NM_019784.5 | NP_062758.3 | 7 - 8 | 1449 | 133 | ||
| GenBank mRNA | AB000682.1 | - | 7 - 8 | 1446 | 133 | |
| AK016434.1 | - | 7 - 8 | 1481 | 133 | ||
| AK016448.1 | - | 7 - 8 | 1439 | 133 | ||
| AK016471.1 | - | 7 - 8 | 1420 | 133 | ||
| BC100432.1 | - | 7 - 8 | 1449 | 30917866 | 133 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tex21
Genomic Map