Assay Details
| Gene Symbol: Hsdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175185.4 | NP_780394.1 | 3 - 4 | 777 | 58 | |
| GenBank mRNA | AK088412.1 | - | 3 - 4 | 761 | 58 | |
| AK150638.1 | - | 1 - 2 | 444 | 58 | ||
| AK152392.1 | - | 1 - 2 | 444 | 58 | ||
| BC065074.1 | - | 3 - 4 | 693 | 6809862 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hsdl1
Genomic Map