Assay Details
| Gene Symbol: Celf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175235.3 | NP_780444.2 | 5 - 6 | 642 | 77 | |
| GenBank mRNA | AK018246.1 | - | 4 - 5 | 439 | 77 | |
| AK034216.1 | - | 4 - 5 | 438 | 77 | ||
| BC052406.1 | - | 5 - 6 | 642 | 5716119 | 77 | |
| BC057083.1 | - | 4 - 5 | 424 | 5707546 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Celf6
Genomic Map